Anti-FOS Goat Antibody

Supplier: OriGene
ORIGTA311326EA 637 CHF
ORIGTA311326
Anti-FOS Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: FOS (FBJ murine osteosarcoma viral oncogene homolog)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR