Anti-MAP3K6 Goat Antibody

Lieferant: OriGene
ORIGTA311308EA 637 CHF
ORIGTA311308
Anti-MAP3K6 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: MAP3K6 (mitogen-activated protein kinase kinase kinase 6)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR