Anti-DAG1 Goat Antibody

Supplier: OriGene
ORIGTA311294EA 637 CHF
ORIGTA311294
Anti-DAG1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.2-0.6?g/ml

Type: Primary
Antigen: DAG1 (dystroglycan 1 (dystrophin-associated glycoprotein 1))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR