Anti-ARHGDIA Goat Antibody

Lieferant: OriGene
ORIGTA311290EA 637 CHF
ORIGTA311290
Anti-ARHGDIA Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: ARHGDIA (Rho GDP dissociation inhibitor (GDI) alpha)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR