Anti-S1PR2 Goat Antibody

Supplier: OriGene
ORIGTA311287EA 637 CHF
ORIGTA311287
Anti-S1PR2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-1.5?g/ml

Type: Primary
Antigen: S1PR2 (sphingosine-1-phosphate receptor 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR