Anti-EFNB2 Goat Antibody

Supplier: OriGene
ORIGTA311283EA 624 CHF
ORIGTA311283
Anti-EFNB2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: EFNB2 (ephrin-B2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR