Anti-SEMA5A Goat Antibody

Supplier: OriGene
ORIGTA311279EA 624 CHF
ORIGTA311279
Anti-SEMA5A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.03?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: SEMA5A (sema domain, seven thrombospondin repeats (type 1 and type 1-like), transmembrane domain (TM) and short cytoplasmic domain, (semaphorin) 5A)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR