Anti-SNAP23 Goat Antibody

Supplier: OriGene
ORIGTA311277EA 637 CHF
ORIGTA311277
Anti-SNAP23 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.2-0.6?g/ml

Type: Primary
Antigen: SNAP23 (synaptosomal-associated protein, 23kDa)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR