Anti-CRISP2 Goat Antibody

Lieferant: OriGene
ORIGTA311263EA 637 CHF
ORIGTA311263
Anti-CRISP2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.05-0.1?g/ml

Type: Primary
Antigen: CRISP2 (cysteine-rich secretory protein 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR