Anti-SOX11 Goat Antibody

Lieferant: OriGene
ORIGTA311256EA 640 CHF
ORIGTA311256
Anti-SOX11 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: SOX11 (SRY (sex determining region Y)-box 11)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR