Anti-NDUFS8 Goat Antibody

Supplier: OriGene
ORIGTA311237EA 624 CHF
ORIGTA311237
Anti-NDUFS8 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: NDUFS8 (NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR