Anti-IL17A Goat Antibody

Supplier: OriGene
ORIGTA311224EA 624 CHF
ORIGTA311224
Anti-IL17A Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: IL17A (interleukin 17A)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR