Anti-ETFB Goat Antibody

Supplier: OriGene
ORIGTA311218EA 637 CHF
ORIGTA311218
Anti-ETFB Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:8000

Type: Primary
Antigen: ETFB (electron-transfer-flavoprotein, beta polypeptide)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR