Anti-CIRBP Goat Antibody

Lieferant: OriGene
ORIGTA311205EA 637 CHF
ORIGTA311205
Anti-CIRBP Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.01-0.03?g/ml; Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: CIRBP (cold inducible RNA binding protein)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR