Anti-CASP12 Goat Antibody

Lieferant: OriGene
ORIGTA311202EA 637 CHF
ORIGTA311202
Anti-CASP12 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: CASP12 (caspase 12 (gene/pseudogene))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR