Anti-MOB2 Goat Antibody

Supplier: OriGene
ORIGTA311199EA 640 CHF
ORIGTA311199
Anti-MOB2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.02-0.06?g/ml; Immunochemistry: 5-10?g/ml

Type: Primary
Antigen: MOB2 (MOB kinase activator 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR