Anti-MECOM Goat Antibody

Supplier: OriGene
ORIGTA311184EA 624 CHF
ORIGTA311184
Anti-MECOM Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: MECOM (MDS1 and EVI1 complex locus)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR