Anti-PCDH17 Goat Antibody

Lieferant: OriGene
ORIGTA311166EA 637 CHF
ORIGTA311166
Anti-PCDH17 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml

Type: Primary
Antigen: PCDH17 (protocadherin 17)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR