Anti-GSTA5 Goat Antibody

Supplier: OriGene
ORIGTA311159EA 627 CHF
ORIGTA311159
Anti-GSTA5 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.3-1?g/ml

Type: Primary
Antigen: GSTA5 (glutathione S-transferase alpha 5)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR