Anti-C12orf29 Goat Antibody

Supplier: OriGene
ORIGTA311158EA 624 CHF
ORIGTA311158
Anti-C12orf29 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: C12orf29 (chromosome 12 open reading frame 29)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR