Anti-HTR7 Goat Antibody

Supplier: OriGene
ORIGTA311145EA 627 CHF
ORIGTA311145
Anti-HTR7 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: HTR7 (5-hydroxytryptamine (serotonin) receptor 7, adenylate cyclase-coupled)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR