Anti-CACNB2 Goat Antibody

Supplier: OriGene
ORIGTA311140EA 637 CHF
ORIGTA311140
Anti-CACNB2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: CACNB2 (calcium channel, voltage-dependent, beta 2 subunit)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR