Anti-ALDH9A1 Goat Antibody

Lieferant: OriGene
ORIGTA311138EA 637 CHF
ORIGTA311138
Anti-ALDH9A1 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.3-1?g/ml; Immunochemistry: 3-5?g/ml; ELISA: 1:16000

Type: Primary
Antigen: ALDH9A1 (aldehyde dehydrogenase 9 family, member A1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR