Anti-ALDH2 Goat Antibody

Lieferant: OriGene
ORIGTA311134EA 637 CHF
ORIGTA311134
Anti-ALDH2 Goat Antibody
Antikörper
Recommended Dilutions: Western Blot: 0.03-0.1?g/ml; ELISA: 1-2?g/ml

Type: Primary
Antigen: ALDH2 (aldehyde dehydrogenase 2 family (mitochondrial))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR