Anti-GNAS Goat Antibody

Supplier: OriGene
ORIGTA311129EA 637 CHF
ORIGTA311129
Anti-GNAS Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: GNAS (GNAS complex locus)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR