Anti-NOTCH3 Goat Antibody

Supplier: OriGene
ORIGTA311128EA 624 CHF
ORIGTA311128
Anti-NOTCH3 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1?g/ml; Immunochemistry: 4-6?g/ml

Type: Primary
Antigen: NOTCH3 (notch 3)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR