Anti-SGCD Goat Antibody

Supplier: OriGene
ORIGTA311125EA 624 CHF
ORIGTA311125
Anti-SGCD Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.1-0.3?g/ml; Immunochemistry: 3-5?g/ml

Type: Primary
Antigen: SGCD (sarcoglycan, delta (35kDa dystrophin-associated glycoprotein))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR