Anti-PEX2 Goat Antibody

Supplier: OriGene
ORIGTA311124EA 624 CHF
ORIGTA311124
Anti-PEX2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PEX2 (peroxisomal biogenesis factor 2)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR