Anti-TLN2 Rabbit Monoclonal Antibody [clone: EPR2536(2)]

Supplier: OriGene
ORIGTA311091EA 872 CHF
ORIGTA311091
Anti-TLN2 Rabbit Monoclonal Antibody [clone: EPR2536(2)]
Antibodies
Recommended Dilutions: Western Blot: 1:50000-1:200000

Type: Primary
Antigen: TLN2 (talin 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR