Anti-STC1 Rabbit Monoclonal Antibody [clone: EPR6055]

Supplier: OriGene
ORIGTA311078EA 932 CHF
ORIGTA311078
Anti-STC1 Rabbit Monoclonal Antibody [clone: EPR6055]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100

Type: Primary
Antigen: STC1 (stanniocalcin 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR