Anti-ITGAV Rabbit Monoclonal Antibody [clone: EPR5583]

Supplier: OriGene
ORIGTA310958EA 962 CHF
ORIGTA310958
Anti-ITGAV Rabbit Monoclonal Antibody [clone: EPR5583]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Flow Cytometry: 1:10-1:100

Type: Primary
Antigen: ITGAV (integrin, alpha V)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR