Anti-POT1 Rabbit Monoclonal Antibody [clone: EPR6319]

Supplier: OriGene
ORIGTA310771EA 997 CHF
ORIGTA310771
Anti-POT1 Rabbit Monoclonal Antibody [clone: EPR6319]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000

Type: Primary
Antigen: POT1 (protection of telomeres 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR