Anti-ABCD3 Rabbit Monoclonal Antibody [clone: EPR5614]

Supplier: OriGene
ORIGTA310768EA 962 CHF
ORIGTA310768
Anti-ABCD3 Rabbit Monoclonal Antibody [clone: EPR5614]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:100-1:250; Flow Cytometry: 1:100-1:500

Type: Primary
Antigen: ABCD3 (ATP-binding cassette, sub-family D (ALD), member 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR