Anti-MGEA5 Rabbit Monoclonal Antibody [clone: EPR7154(B)]

Supplier: OriGene
ORIGTA310690EA 967 CHF
ORIGTA310690
Anti-MGEA5 Rabbit Monoclonal Antibody [clone: EPR7154(B)]
Antibodies
Recommended Dilutions: Western Blot: 1:10000-1:50000; Immunoprecipitation: 1:100-1:500; Immunochemistry: 1:100-1:250

Type: Primary
Antigen: MGEA5 (meningioma expressed antigen 5 (hyaluronidase))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR