Anti-KLKB1 Rabbit Monoclonal Antibody [clone: EPR5707]

Supplier: OriGene
ORIGTA310647EA 961 CHF
ORIGTA310647
Anti-KLKB1 Rabbit Monoclonal Antibody [clone: EPR5707]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1:10-1:100; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: KLKB1 (kallikrein B, plasma (Fletcher factor) 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR