Anti-IL11RA Rabbit Monoclonal Antibody [clone: EPR5446]

Supplier: OriGene
ORIGTA310615EA 780 CHF
ORIGTA310615
Anti-IL11RA Rabbit Monoclonal Antibody [clone: EPR5446]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:500-1:1000; Flow Cytometry: 1:100-1:1000; Immunofluorescence: 1:250-1:500

Type: Primary
Antigen: IL11RA (interleukin 11 receptor, alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR