Anti-ATF3 Rabbit Monoclonal Antibody [clone: EPMIL41]

Supplier: OriGene
ORIGTA310201EA 932 CHF
ORIGTA310201
Anti-ATF3 Rabbit Monoclonal Antibody [clone: EPMIL41]
Antibodies
Recommended Dilutions: Western Blot: 1:50000-1:100000

Type: Primary
Antigen: ATF3 (activating transcription factor 3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR