Anti-AMBP Rabbit Monoclonal Antibody [clone: EPR5880]

Supplier: OriGene
ORIGTA310185EA 823 CHF
ORIGTA310185
Anti-AMBP Rabbit Monoclonal Antibody [clone: EPR5880]
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: AMBP (alpha-1-microglobulin/bikunin precursor)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR