Mouse IgG Isotype Control [clone: 8H10]

Supplier: OriGene
ORIGTA310155EA 616 CHF
ORIGTA310155
Mouse IgG Isotype Control [clone: 8H10]
Antibodies
Recommended Dilutions:

Type: Control
Antigen: ACTB (actin, beta)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR