Anti-ANXA10 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310145EA 456 CHF
ORIGTA310145
Anti-ANXA10 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:200

Type: Primary
Antigen: ANXA10 (annexin A10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR