Anti-ABCB9 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310140EA 381 CHF
ORIGTA310140
Anti-ABCB9 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: ABCB9 (ATP-binding cassette, sub-family B (MDR/TAP), member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR