Anti-THBS1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310133EA 439 CHF
ORIGTA310133
Anti-THBS1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:50-200; ELISA: 1:40000-80000

Type: Primary
Antigen: THBS1 (thrombospondin 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR