Anti-RPL13 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310128EA 373 CHF
ORIGTA310128
Anti-RPL13 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: RPL13 (ribosomal protein L13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR