Anti-PIWIL3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310123EA 466 CHF
ORIGTA310123
Anti-PIWIL3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:100-500; ELISA: 1:40000-80000

Type: Primary
Antigen: PIWIL3 (piwi-like RNA-mediated gene silencing 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR