Anti-PIWIL2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310122EA 448 CHF
ORIGTA310122
Anti-PIWIL2 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:400

Type: Primary
Antigen: PIWIL2 (piwi-like RNA-mediated gene silencing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR