Anti-LOXL1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310116EA 448 CHF
ORIGTA310116
Anti-LOXL1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: LOXL1 (lysyl oxidase-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR