Anti-HCLS1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310115EA 448 CHF
ORIGTA310115
Anti-HCLS1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:50-200; ELISA: 1:40000-80000

Type: Primary
Antigen: HCLS1 (hematopoietic cell-specific Lyn substrate 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR