Anti-DGCR8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310111EA 418 CHF
ORIGTA310111
Anti-DGCR8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:250

Type: Primary
Antigen: DGCR8 (DGCR8 microprocessor complex subunit)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR