Anti-CNTF Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA310109EA 466 CHF
ORIGTA310109
Anti-CNTF Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CNTF (ciliary neurotrophic factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR