Anti-BCL3 Rabbit Polyclonal Antibody

Lieferant: OriGene
ORIGTA310105EA 381 CHF
ORIGTA310105
Anti-BCL3 Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: BCL3 (B-cell CLL/lymphoma 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR